Learn More
Med Vet International Sharps Container, 2 Gallons, Locking Top Flap, Polypropylene, 10/Box
Catalog No. 501180491
Encompass_Preferred
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs. Never recycle the container. Sharps should never be thrown loosely into the trash or toilet. Even Sharps that retract after use, or are very small, should be disposed of like all other sharps. Go to Safeneedledisposal.org for more information about your states regulations. 10 containers per box.
Catalog No. 50-118-0491 Supplier Med Vet International Supplier No. SHARP2GX10
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs. Never recycle the container. Sharps should never be thrown loosely into the trash or toilet. Even Sharps that retract after use, or are very small, should be disposed of like all other sharps. Go to Safeneedledisposal.org for more information about your states regulations. 10 containers per box.