Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sterilis Solutions Sharps Container, 4 Gallon Cylinder
SDP

Catalog No. 23777027
Change view
Click to view available options
Quantity:
10 Pk.
20 Pk.
40 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-777-027 20 Pk.
23-777-026 10 Pk.
23-777-025 40 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-777-027 Supplier Sterilis Solutions Supplier No. 3823310002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterilis Solutions Sharps Container, 4 Gallon Cylinder is a puncture-resistant sharps container with wide circular opening for easy loading of needles.

  • General-purpose sharps container with clear locking lid and marked with biohazard label
  • Compliant with FDA and OSHA standards for sharps and biohazard waste collection
  • Puncture-resistant with wide circular opening for vertical or horizontal loading of needles
  • Made from transparent red puncture-resistant polypropylene that can be autoclaved or incinerated
  • Rotating handle for easy transport and handling
  • Nests to conserve storage space

Specifications

Capacity (Metric) 15 L
Capacity (English) 4 gal.
Color Translucent Red
Dimensions (L x W) 11.37 x 13 in.
Disposable Single-use
Height (Metric) 330.1 mm
Latex Free Yes
Quantity 20 Pk.
Shape Cylindrical
Stackable Yes
Sterility Sterile
Vent Yes
Autoclavable Autoclavable
Height (English) 13 in.
Material Polyethylene
Product Type Sharps Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.