Learn More
WCM INC Isolyser/SMS2400-Sharps Management System. .8 Gallon Sharps disposal
Catalog No. NC2541267
Encompass_Preferred
Isolyser/SMS2400- .8 Gallon container. For Medical Waste Sharps Disposal. Simply fill your Isolyser/SMS container with sharps (needles, blades, endo files, empty carpules and any other potential sharps). When it’s full, add ‘catalyst’ packs to the container, causing the liquid to heat and convert into solid plastic polymer. Once solidified, the sharps are treated; safe for disposal in the regular trash. It’s that easy! All you need for proper Sharps disposal is Isolyser, no other service necessary.
Catalog No. NC2541267 Supplier WCM INC Supplier No. SMS2400
Isolyser/SMS2400- .8 Gallon container. For Medical Waste Sharps Disposal. Simply fill your Isolyser/SMS container with sharps (needles, blades, endo files, empty carpules and any other potential sharps). When it’s full, add ‘catalyst’ packs to the container, causing the liquid to heat and convert into solid plastic polymer. Once solidified, the sharps are treated; safe for disposal in the regular trash. It’s that easy! All you need for proper Sharps disposal is Isolyser, no other service necessary.