Learn More
Oak Ridge Sharps Container with Slide Lid
Catalog No. 22730449
Change view
Click to view available options
Capacity (Metric):
3.7 L
11.3 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-730-449 11.3 L
22-730-446 3.7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-730-449 Supplier Oak Ridge Supplier No. 03301500
Add to Cart
Add to Cart
Designed for by professional health caregivers
  • For use by professional health caregivers
  • Puncture-resistant, transparent body allows for easy viewing
Specifications
Capacity (Metric) 11.3 L
Closure Type Slide Top
Capacity (English) 3 gal.
Material Polypropylene
Product Type Sharps Container