Learn More
Sklar Instruments Sharps Container, Sklar, Sharps Container High Volume Size Red With Locking Rotor Two Gallon, Polypropylene. EA/1.
SDP
Supplier:  Sklar Instruments 101271
Encompass_Preferred
Sharps Container, Sklar, Sharps Container High Volume Size Red With Locking Rotor Two Gallon, Polypropylene. Sklar Sharps Containers provide a safe, readily identifiable canister for various sizes of used needles. They are available in several sizes to accommodate varying quantities of sharps depending on the setting and frequency of sharps usage. This product is a high volume, 2 gallon container. It features a locking rotor for placement of sharps and ensuring safe containment.
Catalog No. 50-143-6952