Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sterilis Solutions Sharps Container Wall Mounting
SDP

Catalog No. 23777034
Change view
Click to view available options
For Use With (Equipment):
2 Gallon Sharps Container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-777-034 2 Gallon Sharps Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-777-034 Supplier Sterilis Solutions Supplier No. 38233441
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wall Mount fixture for Sterilis Solutions 2 Gallon Sharps container with Locking Mechanism.

  • For Sterilis Solutions 2 Gallon Sharps Container
  • With Locking Mechanism

Specifications

Capacity (English) 2 gal.
Color White
Disposable Reusable
Latex Free Yes
Sterility Non-sterile
For Use With (Equipment) 2 Gallon Sharps Container
Mounting Type Wall
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.