Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sharps Containers

Catalog No. 12500102 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
1 qt.
5 qt.
Dimensions (L x W x H):
10.6 x 4.6 x 11.4 in.
4.3 x 3.5 x 6.9 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W x H)
12-500-102 1 qt. 4.3 x 3.5 x 6.9 in.
12-500-100 5 qt. 10.6 x 4.6 x 11.4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-500-102

Designed to fit into blood drawing trays for point-of-use disposal

  • Meet the requirements of BSE/TSE
  • Manufactured under the ISO 13485 standard
  • Puncture-Proof and Durable Construction
  • Meets Medical Biohazard Waste Regulations

Specifications

Certifications/Compliance BSE/TSE
Capacity (English) 1 qt.
Dimensions (L x W x H) 4.3 x 3.5 x 6.9 in.
Height (Metric) 17.4 cm
Length (Metric) 10.8 cm
Width (Metric) 8.8 cm
Width (English) 3.5 in.
Length (English) 4.3 in.
Height (English) 6.9 in.
Material Polypropylene
Product Type Sharps Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.