Learn More
Stoelting™ Sharps Disposable Container
SDP
Catalog No. 10000856 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
1 gal.
Catalog No. Size
10-000-856 1 gal.
1 options
Catalog No. 10-000-856 Supplier Stoelting™ Supplier No. 52190
Add to Cart
Add to Cart
Stoelting™ Sharps Disposable Containers feature one-hand, one-step, no-touch sharps depositing.
  • Containers feature horizontal loading, allowing for maximum syringe capacity
  • Non-mechanical sharps openings minimize tampering and contamination
  • Safe to autoclave or incinerate
Specifications
For Use With (Application) Neuroscience
Size 1 gal.