Learn More
WCM INC Isolyser®/SMS®4000- 1.5G container. Medical Waste Sharps Disposal. Fill with Sharps, Treat on site and throw away in trash.
Catalog No. NC3790351
Encompass_Preferred
Isolyser®/SMS®4000- 1.5 Gallon container. 4 Units in Case. Medical Waste Sharps Disposal. Simply fill your Isolyser/SMS container with sharps (needles, blades, endo files, empty carpules and any other potential sharps). When it’s full, add ‘catalyst’ packs to the container, causing the liquid to heat and convert into solid plastic polymer. Once solidified, the sharps are treated; safe for disposal in the regular trash. It’s that easy! All you need for proper Sharps disposal is Isolyser, no other service necessary.
This item is not returnable. View return policy
Catalog No. NC3790351 Supplier WCM INC Supplier No. SMS4000
Isolyser®/SMS®4000- 1.5 Gallon container. 4 Units in Case. Medical Waste Sharps Disposal. Simply fill your Isolyser/SMS container with sharps (needles, blades, endo files, empty carpules and any other potential sharps). When it’s full, add ‘catalyst’ packs to the container, causing the liquid to heat and convert into solid plastic polymer. Once solidified, the sharps are treated; safe for disposal in the regular trash. It’s that easy! All you need for proper Sharps disposal is Isolyser, no other service necessary.