Learn More
MSC ASI-American Specialties Sharps Disposal Cabinet, Stainless Steel, Surface Mounted
Catalog No. 502837225
Encompass
This sharps disposal cabinet is a surface-mounted unit constructed from durable Type 304 stainless steel with a satin finish. Designed for medical environments, it features a secure, tumbler-locked door and accommodates a specific model of sharps collector (not included). Its robust construction ensures long-lasting use and easy maintenance.
  • Constructed from durable and easy-to-clean stainless steel.
  • Features adjustable shelves for flexible storage and use.
  • Door is secured with a tumbler lock.
  • Marked with a graphic label indicating biohazard.
  • Designed for surface mounting with a stainless steel collar.
  • Accommodates one sharps collector.
Catalog No. 50-283-7225 Supplier MSC Supplier No. 47849385
This sharps disposal cabinet is a surface-mounted unit constructed from durable Type 304 stainless steel with a satin finish. Designed for medical environments, it features a secure, tumbler-locked door and accommodates a specific model of sharps collector (not included). Its robust construction ensures long-lasting use and easy maintenance.
  • Constructed from durable and easy-to-clean stainless steel.
  • Features adjustable shelves for flexible storage and use.
  • Door is secured with a tumbler lock.
  • Marked with a graphic label indicating biohazard.
  • Designed for surface mounting with a stainless steel collar.
  • Accommodates one sharps collector.