Learn More
Research Products International Corp Sharps-Tainer, Small, 5 qt, 16/CS
SDP
Catalog No. 501977820
Encompass
Designed for use in research labs, patient rooms, physicians' offices, clinics and other high-traffic areas. Hinged lid accommodates large items and horizontal drop allows maximum capacity of sharps. The translucent containers and lids help identify fill level. Sharps-Tainers lock for final disposal and can accommodate syringes up to 60cc. Multilingual labeling ensures user safety. Container can be autoclaved or incinerated for final disposal.
Catalog No. 50-197-7820 Supplier Research Products International Corp Supplier No. 108184
Designed for use in research labs, patient rooms, physicians' offices, clinics and other high-traffic areas. Hinged lid accommodates large items and horizontal drop allows maximum capacity of sharps. The translucent containers and lids help identify fill level. Sharps-Tainers lock for final disposal and can accommodate syringes up to 60cc. Multilingual labeling ensures user safety. Container can be autoclaved or incinerated for final disposal.