Learn More
Med Vet International Medegen Sharps Container, 3 Gallon, Red/ Black, Large Tortuous Path Lid, 12.5in x 13.5in x 6in, 12/Case
Supplier:  Med Vet International 8710CS
Encompass_Preferred
Disposal container with translucent top. Contents are always safely enclosed with locking door. Autoclavable polypropylene construction. Built-in phlebotomy needle port
Catalog No. 50-255-9618