Learn More
Med Vet International Medegen Sharps Container, Red Base 10.5in X 3.5in X 10.5in, Horizontal Entry, 3/Pack
Supplier:  Med Vet International 8708X3
Encompass_Preferred
Stackable Biohazard Labeling, Biohazard Symbol Autoclave/Incinerator Safe up to a max of 280F (137C)
Catalog No. 50-255-9620