Learn More
Medchemexpress LLC Shatavarin IV | 84633-34-1 | 99.8% | 887.06 | 1 MG
SDP
Catalog No. 5000426903
Encompass_Preferred
Shatavarin IV (also known as Asparanin B) is a steroidal saponin isolated from the roots of Asparagus racemosus (Liliaceae). It exhibits anticancer activity and has been observed to extend lifespan and alleviate Parkinsonism in Caenorhabditis elegans.
  • Isolated from natural source, Asparagus racemosus.
  • Demonstrates anticancer activity.
  • Elicits lifespan extension in Caenorhabditis elegans.
  • Alleviates Parkinsonism in Caenorhabditis elegans.
  • Reduces intracellular lipofuscin accumulation in C. elegans at 50 μM.
  • Reduces ROS levels in C. elegans (0-100 μM).
  • Increases mRNA expression of stress responsive genes in C. elegans at 50 μM (sod-1, sod-2, sod-3, gst-4, gst-7, ctl-2).
Catalog No. 50-004-26903 Supplier Medchemexpress LLC Supplier No. HYN88041MG
Shatavarin IV (also known as Asparanin B) is a steroidal saponin isolated from the roots of Asparagus racemosus (Liliaceae). It exhibits anticancer activity and has been observed to extend lifespan and alleviate Parkinsonism in Caenorhabditis elegans.
  • Isolated from natural source, Asparagus racemosus.
  • Demonstrates anticancer activity.
  • Elicits lifespan extension in Caenorhabditis elegans.
  • Alleviates Parkinsonism in Caenorhabditis elegans.
  • Reduces intracellular lipofuscin accumulation in C. elegans at 50 μM.
  • Reduces ROS levels in C. elegans (0-100 μM).
  • Increases mRNA expression of stress responsive genes in C. elegans at 50 μM (sod-1, sod-2, sod-3, gst-4, gst-7, ctl-2).