Learn More
Ergodyne™ SHAX™ 6000 Heavy-Duty Pop-Up Tent
Catalog No. 19819696 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Quantity:
1/Ea.
Catalog No. Color Quantity
19-819-696 Blue 1/Ea.
1 options
Catalog No. 19-819-696 Supplier Ergodyne™ Supplier No. 12905
SHAX™ 6000 Heavy-Duty Pop-Up Tent is a durable, instant shelter that provides fast, long-lasting relief from the sun, wind, and rain.
  • 10 x 10 ft. (3 x 3 m) canopy provides 100 square feet of shaded area
  • Steel frame and reinforced corners on the canopy to withstand tough conditions
  • Rugged 300D polyester canopy with PU coating and UV treatments
  • ABS plastic sliders on legs adjust to 3 height settings
  • Double-vented canopy for maximum airflow
  • Large storage bag with wheels holds collapsed tent with canopy still on
  • 4 heavy-duty ground stakes and 4 ropes included
  • Meets CPAI-84 flame resistance code
Specifications
Material Steel, Polyester, ABS Plastic
Color Blue
Dimensions (L x W) 10 x 10 ft. (3 x 3 m)
Quantity 1/Ea.