Learn More
Ergodyne™ SHAX™ 6100 Lightweight Industrial Umbrella
Catalog No. 19819702 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Lime
Catalog No. Color
19-819-702 Lime
1 options
Catalog No. 19-819-702 Supplier Ergodyne™ Supplier No. 12967
Provides fast and portable relief from the elements to increase worker productivity and reduce weather-related delays.
  • Engineered with a powder-coated steel frame, a reinforced weather-proof canopy and 300 denier polyester material
  • Lightweight
  • Tilt-top feature allows users to shade themselves from ultraviolet (UV) sun rays any hour of the day.
  • Vented lime top provides airflow and visibility
  • Ideal construction umbrella for hot jobsites
  • Polyester shell also has protective coatings (Polyurethane (PU) and UV) which provide the ultimate shelter from sun/UV, rain, and wind
  • Helps prevent heat-related illnesses on any residential, commercial, agricultural, construction or highway setting
Specifications
Color Lime
Diameter (English) 7 ft.
Diameter (Metric) 2.1 m
Quantity 1/Cs.