Learn More
Aviva Systems Biology Corp SHB Antibody - N-terminal region (ARP58762_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58762_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SHB. Alternate names: RP11-3J10.8, bA3J10.2ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62414