Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6462MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM605. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Liver, Placenta or Testis. Cellular Staining: Cytoplasmic & Extracellular (Secreted). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human SHBG protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 45kDa.Chromosome: 17p13.1. Gene Symbol: SHBG. Gene ID: 6462. Ensemble: ENSG00000129214. Uniprot: NP_001276044.
Catalog No. 50-188-5164