Learn More
Crystal Chem Inc Sex Hormone Binding Globulin (SHBG) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80978
Encompass
The Sex Hormone Binding Globulin (SHBG) ELISA is used to quantify levels of human SHBG in serum using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. High sensitivity of 0.1 nmol/L. Wide dynamic range of 3.3 - 295 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7880