Learn More
Drg International Inc DRG SHBG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA2996R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of sex hormone binding globulin (SHBG) in serum and plasma (EDTA-, heparin- or citrate plasma). This product is for Research Use Only. The DRG SHBG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site of the SHBG molecule.  An aliquot sample containing endogenous SHBG is incubated in the coated well. 
Catalog No. 50-194-7552