Learn More
Enzo Life Sciences Shc monoclonal antibody (11F6) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905665100
Encompass_Preferred
Clone: 11F6. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of Shc. UniProt: P29353. Species Reactivity: Human, Mouse. Applications: ELISA, ICC, IHC, WB. Recommended dilutions: Immunohistochemistry (1:50). Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-658-420