Learn More
Aviva Systems Biology Corp Shc3 Antibody - N-terminal region (ARP56957_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56957_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Shc3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MGC124163, N-Shc, Rai, ShcCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-244