Learn More
Aviva Systems Biology Corp SHCBP1L Antibody - N-terminal region (ARP53794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHCBP1L. It was validated on Western Blot. . Alternate names: GE36, MGC26911, MGC48296, SHCBP1L, C1orf14ApplicationWB. Species reactivity: Human
Catalog No. 50-285-888