Learn More
Grainger Reed Manufacturing SC125 Scissor Shears, 8.5 in., 1-3/4 in. Capacity, 1 ea.
Catalog No. NC1983937
Encompass_Preferred
The SC125 Scissor Shears are designed for precision cutting of various plastic pipes and tubing materials. Featuring a durable construction and specialized blade, this tool provides reliable performance for plumbing and industrial applications.
  • Features an industrial-grade, non-stick steel blade.
  • Constructed with a lightweight fiberglass body and ribbed handle for a secure grip.
  • Designed for cutting ABS, nylon, PEX, polyethylene, rubber, and thin-wall PVC.
  • Offers a maximum cutting capacity of 1-3/4 in.
  • Includes safety and operational guidelines recommending blade-on-bottom usage.
  • Made in the USA.
Catalog No. NC1983937 Supplier Grainger Supplier No. 38HV24
The SC125 Scissor Shears are designed for precision cutting of various plastic pipes and tubing materials. Featuring a durable construction and specialized blade, this tool provides reliable performance for plumbing and industrial applications.
  • Features an industrial-grade, non-stick steel blade.
  • Constructed with a lightweight fiberglass body and ribbed handle for a secure grip.
  • Designed for cutting ABS, nylon, PEX, polyethylene, rubber, and thin-wall PVC.
  • Offers a maximum cutting capacity of 1-3/4 in.
  • Includes safety and operational guidelines recommending blade-on-bottom usage.
  • Made in the USA.