Learn More
Med Vet International Dynarex Bandage Shears, 7.5", Yellow, 200/Case
Catalog No. 502830472
Encompass_Preferred
Dynarex DynaSafety Bandage Shears are high-quality rescue tools designed to cut through gauze, tape, and clothing. The reliable Bandage Shears are crafted with hardened stainless-steel blades and feature a serrated edge strong enough to cut through seatbelts and other thick materials. The premium Shears feature contoured handles for improved grip and easy handling. Dynarex Bandage Shears are available in a variety of sizes and colors for easy identification. Dynarex DynaSafety Bandage Shears are designed with premium stainless-steel blades and a sharpened serrated edge to cut through gauze, tapes, clothing, and seatbelts. The high-quality shears feature contoured handles for a sturdy, reliable grip.
Catalog No. 50-283-0472 Supplier Med Vet International Supplier No. 4187
Dynarex DynaSafety Bandage Shears are high-quality rescue tools designed to cut through gauze, tape, and clothing. The reliable Bandage Shears are crafted with hardened stainless-steel blades and feature a serrated edge strong enough to cut through seatbelts and other thick materials. The premium Shears feature contoured handles for improved grip and easy handling. Dynarex Bandage Shears are available in a variety of sizes and colors for easy identification. Dynarex DynaSafety Bandage Shears are designed with premium stainless-steel blades and a sharpened serrated edge to cut through gauze, tapes, clothing, and seatbelts. The high-quality shears feature contoured handles for a sturdy, reliable grip.