Learn More
Leinco Technologies Inc 1X ClearFlow™ Sheath Fluid | 20L
SDP
Catalog No. NC0834401
Encompass_Preferred
ClearFlow™ Sheath Fluid is specifically formulated isotonic solution for transporting particles through a flow cytometer and supplied "Ready to Use". ClearFlow™ is specially manufactured in our ISO 7(class 10,000) cleanroom suite for low particulate count (SEE ABOVE EXPERIMENT) and fluorescence background to guarantee better signal to noise ratio measurements.
Catalog No. NC0834401 Supplier Leinco Technologies Inc Supplier No. S621
ClearFlow™ Sheath Fluid is specifically formulated isotonic solution for transporting particles through a flow cytometer and supplied "Ready to Use". ClearFlow™ is specially manufactured in our ISO 7(class 10,000) cleanroom suite for low particulate count (SEE ABOVE EXPERIMENT) and fluorescence background to guarantee better signal to noise ratio measurements.