Learn More
Bio Rad Laboratories SHEATH FLUID BOTTLE
Catalog No. 509013080
Encompass
Microplate Instrumentation Accessories; Sheath Fluid Bottle; Bio-Rad; Polypropylene; With sheath-fluid and air-intake ports and tubing; For use with the Bio-Plex 200 system; 1L
Catalog No. 50-901-3080 Supplier Bio Rad Laboratories Supplier No. 171002010
Microplate Instrumentation Accessories; Sheath Fluid Bottle; Bio-Rad; Polypropylene; With sheath-fluid and air-intake ports and tubing; For use with the Bio-Plex 200 system; 1L