Learn More
Med Vet International Sheather’s Sugar Flotation Solution, 500mL, J1028, Each
Catalog No. 5000583181
Encompass_Preferred
Find more eggs!! Recent studies have proven that centrifugation of fecal samples is for more effective than traditional standing passive flotation. Furthermore, a flotation media must have a higher specific gravity than 1.20 to float many of the common parasite ova. Sheather’s sugar solution has a specific gravity of 1.27 which is high enough to float any ova.Basic Physics: If an ova has a higher specific gravity than the flotation solution, it will not float nor will it be detected.
Catalog No. 50-005-83181 Supplier Med Vet International Supplier No. JOR1028
Find more eggs!! Recent studies have proven that centrifugation of fecal samples is for more effective than traditional standing passive flotation. Furthermore, a flotation media must have a higher specific gravity than 1.20 to float many of the common parasite ova. Sheather’s sugar solution has a specific gravity of 1.27 which is high enough to float any ova.Basic Physics: If an ova has a higher specific gravity than the flotation solution, it will not float nor will it be detected.