Learn More
Med Vet International Sheather's Sugar Fecal Floatation Solution, Gallon
Catalog No. 502232659
Encompass_Preferred
The only Sheather's solution made with natural preservative...No Formaldehyde in thei formula!! Find more eggs!! Recent studies have proven that centrifugation of fecal samples is for more effective than traditional standing passive flotation. Furthermore, a flotation media must have a higher specific gravity than 1.20 to float many of the common parasite ova. Sheather’s sugar solution has a specific gravity of 1.27 which is high enough to float any ova. Basic Physics: If an ova has a higher specific gravity than the flotation solution, it will not float nor will it be detected.Products are not always manufacturer specific. Actual product appearance may differ from website image.
This item is not returnable. View return policy
Catalog No. 50-223-2659 Supplier Med Vet International Supplier No. PHSHEATHERS
The only Sheather's solution made with natural preservative...No Formaldehyde in thei formula!! Find more eggs!! Recent studies have proven that centrifugation of fecal samples is for more effective than traditional standing passive flotation. Furthermore, a flotation media must have a higher specific gravity than 1.20 to float many of the common parasite ova. Sheather’s sugar solution has a specific gravity of 1.27 which is high enough to float any ova. Basic Physics: If an ova has a higher specific gravity than the flotation solution, it will not float nor will it be detected.Products are not always manufacturer specific. Actual product appearance may differ from website image.