Learn More
American Research Products Inc Sheep 5 hydroXytryptamine sero
Catalog No. 5000330071
Encompass
Sheep 5-hydroXytryptamine/serotonin (5HT/ST) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-10 ng/mL Sensitivity 0 312 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-30071 Supplier American Research Products Inc Supplier No. CSBE14929SH
Sheep 5-hydroXytryptamine/serotonin (5HT/ST) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-10 ng/mL Sensitivity 0 312 ng/mL Sample serum plasma Method Competitive