Learn More
American Research Products Inc Sheep adrenocorticotropic horm
Catalog No. 5000326980
Encompass
Sheep adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 0 097 pg/mL-400 pg/mL Sensitivity 0 097 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26980 Supplier American Research Products Inc Supplier No. CSBEQ027618SH
Sheep adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 0 097 pg/mL-400 pg/mL Sensitivity 0 097 pg/mL Sample serum plasma Method Competitive