Learn More
Innovative Research Inc Sheep Anti Human Factor VIII Polyclonal Fractionated 5mg
Catalog No. 502035974
Encompass_Preferred
Sheep Anti Human Factor VIII Polyclonal Fractionated from Innovative Research is a polyclonal antibody in frozen liquid format, buffered in 50% Glycerol/H2O (vol/vol), with a concentration of 7mg/ml.
Catalog No. 50-203-5974 Supplier Innovative Research Inc Supplier No. ISHAHUFVIIIGF5MG
Sheep Anti Human Factor VIII Polyclonal Fractionated from Innovative Research is a polyclonal antibody in frozen liquid format, buffered in 50% Glycerol/H2O (vol/vol), with a concentration of 7mg/ml.