Learn More
Immunology Consultants anti-Goat IgG h+l Antibody (Sheep) - Affinity Purified
SDP
Catalog No. 502248876
Encompass_Preferred
This affinity‑purified sheep anti‑goat IgG (H+L) antibody provides broad, high‑affinity recognition of both heavy and light chains of goat IgG. Suitable for ELISA, Western blotting, immunoprecipitation, and secondary detection workflows, it delivers strong signal and low background. Produced by Immunology Consultants, this 1 mg reagent offers consistent performance across diverse immunoassay formats, making it a versatile secondary antibody for goat‑derived primary antibodies.
This item is not returnable. View return policy
Catalog No. 50-224-8876 Supplier Immunology Consultants Supplier No. SGHL50A
This affinity‑purified sheep anti‑goat IgG (H+L) antibody provides broad, high‑affinity recognition of both heavy and light chains of goat IgG. Suitable for ELISA, Western blotting, immunoprecipitation, and secondary detection workflows, it delivers strong signal and low background. Produced by Immunology Consultants, this 1 mg reagent offers consistent performance across diverse immunoassay formats, making it a versatile secondary antibody for goat‑derived primary antibodies.