Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Sheep anti-Human, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP512PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP512PMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP512PMI Supplier MilliporeSigma Supplier No. AP512P

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

Sheep Anti-Human IgM Polyclonal Antibody validated for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sheep anti-Human IgM
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Purified sheep polyclonal in PBS containing 0.01% Thimerisol and 15mg/mL BSA. Depleted against rabbit IgM and human IgG.
Host Species Sheep
Immunogen Human anti-Human IgM (myeloma) Fc5u fragment
Purification Method Depletion
Quantity 500 μg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8 °C protected from light in undiluted aliquots for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.