Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, Biotin-SP Conjugate Sheep anti-Rabbit, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP510BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP510BMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP510BMI Supplier MilliporeSigma Supplier No. AP510B

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

Sheep anti-Rabbit IgG, Biotin-SP Conjugate Antibody is an antibody against Rabbit IgG for use in immunohistochemistry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sheep anti-Rabbit IgG
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1mg/mL
Conjugate Biotin
Formulation Purified sheep polyclonal in PBS containing 15mg/mL BSA and 0.1% sodium azide.
Host Species Sheep
Immunogen Rabbit IgG
Purification Method Octanoic Acid and Ammonium Sulfate cut. Depleted against Goat IgG and Rabbit IgM
Quantity 1 mg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8 °C protected from light in undiluted aliquots for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.