Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sheep anti-Rabbit IgG, Cy5, Polyclonal, Secondary Antibody,

Catalog No. ap510smi Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
Conjugate:
Cyanine5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AP510SMI 500 μg Cyanine5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP510SMI Supplier MilliporeSigma™ Supplier No. AP510S
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Specifically detects Rabbit IgG in Rabbit samples, and it is validated for Immunocytochemistry

Immunoglobulin G (IgG) is part of the family of glycoprotein immunoglobulins which stage immune responses against pathogenic organisms. IgG contributes to the immune response by mobilizing innate immune effector cells and inducing and pro-inflammatory responses. Such proinflammatory responses are mediated by Fcg receptors which interact with sialylated Fc fragments on IgG molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Cyanine5
Formulation Purified sheep antibody Conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
Host Species Sheep
Immunogen Rabbit IgG
Purification Method Protein A Purfied
Quantity 500 μg
Regulatory Status RUO
Research Discipline Secondary and Control Antibodies
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.