Learn More
American Research Products Inc Sheep arachidonic Acid AA EL
Catalog No. 5000327966
Encompass
Sheep arachidonic Acid (AA) ELISA kit from CUSABIO Detection Range 15 625 ng/mL-1000 ng/mL Sensitivity 13 797 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-27966 Supplier American Research Products Inc Supplier No. CSBEQ027590SH
Sheep arachidonic Acid (AA) ELISA kit from CUSABIO Detection Range 15 625 ng/mL-1000 ng/mL Sensitivity 13 797 ng/mL Sample serum plasma Method Competitive