Learn More
Hemostat Laboratories SHEEP BLOOD DEFIB 250ML
SDP
Catalog No. 50863755
Encompass_Preferred
Defibrinated Sheep Blood (250Ml), Aseptically Collected From A Controlled Herd Of Healthy Animals Of U.S. Origin. Mechanically Defibrinated Without Anticoagulants Or Other Additives. Proprietary Screening Process Insures Low Fragility And Consistent Color. For In Vivo Diagnostic Use Only.
Catalog No. 50-863-755 Supplier Hemostat Laboratories Supplier No. DSB250
Defibrinated Sheep Blood (250Ml), Aseptically Collected From A Controlled Herd Of Healthy Animals Of U.S. Origin. Mechanically Defibrinated Without Anticoagulants Or Other Additives. Proprietary Screening Process Insures Low Fragility And Consistent Color. For In Vivo Diagnostic Use Only.