Learn More
Hemostat Laboratories SHEEP BLOOD DEFIB 500ML
SDP
Catalog No. 50863756
Encompass_Preferred
Sheep Blood Defibrinated, 500ml Aseptically collected from healthy animals of U.S. origin. Mechanically defibrinated without anticoagulants or other additives. Proprietary sceening process insures low fragility and consistent color.
Catalog No. 50-863-756 Supplier Hemostat Laboratories Supplier No. DSB500
Sheep Blood Defibrinated, 500ml Aseptically collected from healthy animals of U.S. origin. Mechanically defibrinated without anticoagulants or other additives. Proprietary sceening process insures low fragility and consistent color.