Learn More
Rockland Immunochemicals SHEEP BLOOD Defibrinated - R111-0100
SDP
Catalog No. 501059121
Encompass_Preferred
Defibrinated sheep blood is aseptically collected and prepared to order from our colony of animals. Fibrin is mechanically removed during the clotting process without the presence of anticoagulants or other additives. Defibrinated sheep blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Defibrinated sheep blood is perishable and is collected and processed upon receipt of your order.
Catalog No. 50-105-9121 Supplier Rockland Immunochemicals Supplier No. R1110100
Defibrinated sheep blood is aseptically collected and prepared to order from our colony of animals. Fibrin is mechanically removed during the clotting process without the presence of anticoagulants or other additives. Defibrinated sheep blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Defibrinated sheep blood is perishable and is collected and processed upon receipt of your order.