Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemostat Laboratories Sheep Blood w/ Citrate, 100 ml, Liquid, Aseptically Collected, Anticoagulant: Sodium Citrate, Anticoagulant Ratio: 90/10
SDP

Catalog No. NC1046426
Encompass_Preferred

Hemostat Laboratories SBC100 Sheep Blood with Citrate is a sterile, whole animal blood product prepared for various scientific applications. This product contains aseptically collected sheep blood with sodium citrate as an anticoagulant to prevent clotting. It is ideal for biomedical research, cell culture, diagnostic manufacturing, and medical device development. Due to its nature, the product has a short shelf life and is shipped from a California-based warehouse.

  • Species: Sheep
  • Anticoagulant: Sodium citrate
  • Volume: 100 ml
  • Anticoagulant ratio: 90/10 (blood to citrate)
  • Collection method: Aseptically collected
  • Primary use: Scientific applications
  • Form: Liquid
  • Shelf life: Short
  • Shipment origin: California warehouse

Catalog No. NC1046426 Supplier Hemostat Laboratories Supplier No. SBC100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hemostat Laboratories SBC100 Sheep Blood with Citrate is a sterile, whole animal blood product prepared for various scientific applications. This product contains aseptically collected sheep blood with sodium citrate as an anticoagulant to prevent clotting. It is ideal for biomedical research, cell culture, diagnostic manufacturing, and medical device development. Due to its nature, the product has a short shelf life and is shipped from a California-based warehouse.

  • Species: Sheep
  • Anticoagulant: Sodium citrate
  • Volume: 100 ml
  • Anticoagulant ratio: 90/10 (blood to citrate)
  • Collection method: Aseptically collected
  • Primary use: Scientific applications
  • Form: Liquid
  • Shelf life: Short
  • Shipment origin: California warehouse

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.