Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Preserved Sheep Brain, Dura Mater Intact
SDPSCI_ED

Catalog No. S28065 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S28065 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28065 Supplier Carolina™ Supplier No. 228711
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to study mammalian brain structure.

  • Explores general structures representative of mammals
  • Connects structure to function
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe™ is an odorless fluid designed to minimize the unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 1/Pk.
Packaging Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.