Learn More
Carolina™ Formalin Preserved Sheep Brain, Dura Mater Removed, Hypophysis Intact
SDPSCI_ED
Catalog No. S28071 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1/Pk.
Catalog No. Quantity
S28071 1/Pk.
1 options
Catalog No. S28071 Supplier Carolina™ Supplier No. 228746
Add to Cart
Add to Cart
Used to study mammalian brain structure.
  • Similar to human brain
  • Sheep brain with dura mater removed and hypophysis (pituitary gland) intact to aid in brain study, shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Odorless fluid designed to minimize unpleasant odor of formaldehyde
  • Plain: No color injection
  • Individually damp-packed in vacuum-sealed bags
Specifications
Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 1/Pk.
Packaging Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov