Learn More
American Research Products Inc Sheep Corticoliberin CRH ELI
Catalog No. 5000328044
Encompass
Sheep Corticoliberin (CRH) ELISA kit from CUSABIO Detection Range 14 pg/mL-500 pg/mL Sensitivity 14 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28044 Supplier American Research Products Inc Supplier No. CSBEL005963SH
Sheep Corticoliberin (CRH) ELISA kit from CUSABIO Detection Range 14 pg/mL-500 pg/mL Sensitivity 14 pg/mL Sample serum plasma tissue homogenates Method Competitive