Learn More
Biomatik Corporation Sheep Fibroblast Growth Factor 1, Acidic (FGF1) ELISA Kit
Supplier:  Biomatik Corporation EKU11651
Encompass_Preferred
Sheep Fibroblast Growth Factor 1, Acidic (FGF1) ELISA Kit for detecting FGF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Sheep (Ovis aries). Detection Range: 3.12-200pg/ml. Sensitivity: 1.22pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7M303. Gene ID: 101109353. Target Synonyms: ECGF; AFGF; ECGFA; ECGFB; FGF-Alpha; FGFA; HBGF1; ECGFB; HBGF1; Acidic Fibroblast Growth Factor; heparin-binding growth factor 1; Endothelial Cell Growth Factor, Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8105