Learn More
American Research Products Inc Sheep ghrelin obestatin prepro
Catalog No. 5000326965
Encompass
Sheep ghrelin/obestatin prepropeptide (GHRL) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26965 Supplier American Research Products Inc Supplier No. CSBEL009414SH
Sheep ghrelin/obestatin prepropeptide (GHRL) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich