Learn More
Biomatik Corporation Sheep Hemoglobin (HB) ELISA Kit
Supplier:  Biomatik Corporation EKU11960
Encompass_Preferred
Sheep Hemoglobin (HB) ELISA Kit for detecting HB in serum, plasma, erythrocyte lysates, other biological fluids. Species Reactivity: Sheep (Ovis aries). Detection Range: 6.17-500ug/ml. Sensitivity: 2.63ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P68240; P02075. Target Synonyms: Hgb; Haemoglobin; Heterotetramer (αβ) 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6987