Learn More
Rockland Immunochemicals Sheep IgG F(c) Antibody 2 mg
SDP
Catalog No. 501943935
Encompass_Preferred
Anti-Sheep IgG F(c) generated in rabbit is a proteolytic fragment of immunoglobulin G (IgG) obtained by limited digestion with the enzyme papain under controlled conditions of temperature time and pH. Receptors bind the Fc portion of sheep IgG and often this fragment is removed from immunoglobulins to minimize receptor binding and lower background reactivity. Anti-Sheep IgG F(c) antibody is ideal for investigators in Cancer Immunology and Microbiology research.
Catalog No. 50-194-3935 Supplier Rockland Immunochemicals Supplier No. 6134103
Anti-Sheep IgG F(c) generated in rabbit is a proteolytic fragment of immunoglobulin G (IgG) obtained by limited digestion with the enzyme papain under controlled conditions of temperature time and pH. Receptors bind the Fc portion of sheep IgG and often this fragment is removed from immunoglobulins to minimize receptor binding and lower background reactivity. Anti-Sheep IgG F(c) antibody is ideal for investigators in Cancer Immunology and Microbiology research.