Learn More
Rockland Immunochemicals Sheep IgG (H&L) Antibody Alkaline Phosphatase Conjugated 1 mg
SDP
Catalog No. 501943938
Encompass_Preferred
Anti-Sheep IgG (H&L) antibody generated in donkey detects specifically sheep IgG heavy and light chains (H&L). This Anti sheep antibody is conjugated to alkaline phosphatase and this secondary antibody is suitable for immunoblotting and immunoassays.Anti-Sheep IgG (H&L) antibody is ideal for investigators in Cancer Immunology and Microbiology research.
Catalog No. 50-194-3938 Supplier Rockland Immunochemicals Supplier No. 6137502
Anti-Sheep IgG (H&L) antibody generated in donkey detects specifically sheep IgG heavy and light chains (H&L). This Anti sheep antibody is conjugated to alkaline phosphatase and this secondary antibody is suitable for immunoblotting and immunoassays.Anti-Sheep IgG (H&L) antibody is ideal for investigators in Cancer Immunology and Microbiology research.