Learn More
Biotang Inc Sheep soluble interleukin-1 receptor I,IL-1sR I ELISA Kit, 96T
Catalog No. 50155568 Shop All BIOTANG Inc Products
Encompass_Preferred
Sheep soluble interleukin-1 receptor I,IL-1sR I ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.
Catalog No. 50-155-568 Supplier Biotang Inc Supplier No. SH2824
Sheep soluble interleukin-1 receptor I,IL-1sR I ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.